Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRELD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031717.3 | NP_001026887.1 | 8 | 886 | ||
NM_001077415.2 | NP_001070883.1 | 8 | 886 | |||
NM_015513.4 | NP_056328.2 | 7 | 1386 | |||
XM_011534108.1 | XP_011532410.1 | 8 | 871 | |||
XM_017007175.1 | XP_016862664.1 | 8 | 871 | |||
GenBank mRNA | AF452623.1 | - | 7 | 1147 | ||
AK314113.1 | - | 7 | 1330 | |||
AL050275.1 | - | 7 | 1386 | |||
AY358363.1 | - | 8 | 889 | |||
BC008720.2 | - | 8 | 871 | 2821362 | ||
BF182978.1 | - | 3 | 268 | |||
BX358821.2 | - | 8 | 834 | |||
CR457380.1 | - | 7 | 787 |