Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309192.1 | NP_001296121.1 | 3 | 483 | ||
NM_001309193.1 | NP_001296122.1 | 3 | 483 | |||
NM_004442.7 | NP_004433.2 | 3 | 483 | |||
NM_017449.4 | NP_059145.2 | 3 | 483 | |||
XM_006710441.3 | XP_006710504.1 | 3 | 576 | |||
XM_006710442.3 | XP_006710505.1 | 3 | 551 | |||
GenBank mRNA | AB210018.1 | - | 3 | 392 | ||
AF025304.1 | - | 3 | 363 | |||
AK299701.1 | - | 4 | 422 | |||
BC007903.2 | - | 2 | 261 | |||
BC067861.1 | - | 3 | 482 | |||
D31661.1 | - | 3 | 344 | |||
DB282024.1 | - | 4 | 424 | |||
L36643.1 | - | 3 | 296 | |||
L41939.1 | - | 3 | 442 |