Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170961.1 | NP_001164432.1 | 4 | 562 | ||
NM_001170962.1 | NP_001164433.1 | 3 | 535 | |||
NM_001170963.1 | NP_001164434.1 | 4 | 386 | |||
NM_001555.4 | NP_001546.2 | 4 | 562 | |||
NM_205833.3 | NP_991402.1 | 4 | 562 | |||
XM_011531330.1 | XP_011529632.1 | 5 | 460 | |||
XM_011531333.1 | XP_011529635.1 | 4 | 562 | |||
XM_011531334.1 | XP_011529636.1 | 4 | 562 | |||
GenBank mRNA | AB058894.2 | - | 4 | 414 | ||
AF034198.1 | - | 3 | 384 | |||
AK057916.1 | - | 4 | 393 | |||
AK226008.1 | - | 4 | 419 | |||
AK307871.1 | - | 4 | 438 | |||
BC063884.1 | - | 4 | 398 | 30387876 | ||
Y10523.1 | - | 3 | 359 |