Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039550.1 | NP_001034639.1 | 2 | 253 | ||
NM_006736.5 | NP_006727.2 | 2 | 253 | |||
GenBank mRNA | AK289626.1 | - | 2 | 76 | ||
AK292761.1 | - | 2 | 81 | |||
AK293888.1 | - | 2 | 205 | |||
AK312723.1 | - | 2 | 260 | |||
BC011609.2 | - | 2 | 195 | 3619734 | ||
BC047056.1 | - | 1 | 56 | 5185009 | ||
BT007088.1 | - | 1 | 16 | |||
BX420085.2 | - | 255 | ||||
S37374.1 | - | 1 | 41 | |||
S37375.1 | - | 1 | 41 |