Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 4 | 488 | ||
NM_022559.3 | NP_072053.1 | 4 | 443 | |||
NM_022560.3 | NP_072054.1 | 3 | 368 | |||
GenBank mRNA | A00469.1 | - | 4 | 451 | ||
AV753220.1 | - | 2 | 308 | |||
AY613431.1 | - | 4 | 474 | |||
AY613432.1 | - | 4 | 474 | |||
BC062475.1 | - | 4 | 408 | 30393581 | ||
BC075012.2 | - | 4 | 439 | 30915404 | ||
BC075013.2 | - | 4 | 439 | 30915617 | ||
BC090045.1 | - | 4 | 455 | 30394115 | ||
CD108592.1 | - | 3 | 334 | |||
V00519.1 | - | 4 | 452 |