Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 1 | 960 | ||
NM_205862.1 | NP_995584.1 | 2 | 173 | |||
GenBank mRNA | AY435141.1 | - | 1 | 821 | ||
BC019861.1 | - | 1 | 855 | 4454322 | ||
BM688740.1 | - | 2 | 173 | |||
BM924331.1 | - | 2 | 191 | |||
DQ364250.1 | - | 1 | 829 | |||
J04093.1 | - | 1 | 905 |