Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 9 | 1085 | ||
XM_011531336.1 | XP_011529638.1 | 9 | 1363 | |||
XM_011531337.1 | XP_011529639.1 | 2 | 1392 | |||
XM_017029507.1 | XP_016884996.1 | 8 | 1102 | |||
GenBank mRNA | AB209849.1 | - | 9 | 1040 | ||
AK123515.1 | - | 2 | 1048 | |||
AK296958.1 | - | 4 | 314 | |||
AK313467.1 | - | 9 | 1074 | |||
BC009960.2 | - | 9 | 1072 | 4300487 | ||
BC015768.1 | - | 9 | 1089 | 4868206 | ||
U81379.3 | - | 9 | 1082 | |||
Y09328.1 | - | 9 | 1075 | |||
Y10659.1 | - | 9 | 1085 |