Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 11 | 1248 | ||
XM_011531336.1 | XP_011529638.1 | 11 | 1526 | |||
XM_011531337.1 | XP_011529639.1 | 4 | 1555 | |||
XM_017029507.1 | XP_016884996.1 | 10 | 1265 | |||
GenBank mRNA | AB209849.1 | - | 11 | 1203 | ||
AK123515.1 | - | 4 | 1211 | |||
AK296958.1 | - | 6 | 477 | |||
AK313467.1 | - | 11 | 1237 | |||
BC009960.2 | - | 11 | 1235 | 4300487 | ||
BC015768.1 | - | 11 | 1252 | 4868206 | ||
U81379.3 | - | 11 | 1245 | |||
Y09328.1 | - | 11 | 1238 | |||
Y10659.1 | - | 11 | 1248 |