Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002253.2 | NP_002244.1 | 23 | 3400 | ||
GenBank mRNA | AB209901.1 | - | 23 | 3385 | ||
AF035121.1 | - | 23 | 3401 | |||
AF063658.1 | - | 23 | 3098 | |||
BC131822.1 | - | 23 | 3160 | 40129287 | ||
L04947.1 | - | 23 | 3092 | |||
X61656.1 | - | 23 | 3092 |