Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003505.1 | NP_003496.1 | 1 | 3246 | ||
GenBank mRNA | AB017363.1 | - | 1 | 3246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX870692.1 | - | 1 | 711 | Chr. 7 - 91267003 - 91267807 [+] on Build GRCh38 |
Set Membership: |
G-protein coupled receptor |