Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005738.1 | NP_001005738.1 | 2 | 737 | ||
NM_001462.3 | NP_001453.1 | 2 | 835 | |||
XM_006723120.3 | XP_006723183.1 | 5 | 1568 | |||
XM_017026542.1 | XP_016882031.1 | 4 | 1412 | |||
GenBank mRNA | AF054013.1 | - | 1 | 692 | ||
AK290557.1 | - | 3 | 883 | |||
BC029125.2 | - | 1 | 706 | 5180065 | ||
BC071722.1 | - | 2 | 1007 | 5212947 | ||
BM922150.1 | - | 2 | 773 | |||
D10922.1 | - | 2 | 726 | |||
M76672.1 | - | 1 | 692 | |||
M84562.1 | - | 2 | 1464 | |||
M88107.1 | - | 2 | 731 | |||
U81501.1 | - | 1 | 712 | |||
X63819.1 | - | 2 | 845 |
Set Membership: |
G-protein coupled receptor |