Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036501.1 | 1 | 4241 | |||
GenBank mRNA | AK092714.1 | - | 1 | 2248 | ||
AK127989.1 | - | 1 | 1480 |