Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002030.4 | NP_002021.3 | 2 | 539 | ||
XM_011526687.2 | XP_011524989.1 | 2 | 430 | |||
GenBank mRNA | AK123030.1 | - | 3 | 482 | ||
AK292541.1 | - | 3 | 528 | |||
BC059388.1 | - | 2 | 522 | 30332008 | ||
BC069070.1 | - | 1 | 343 | 7216883 | ||
BC069593.1 | - | 1 | 343 | 7262465 | ||
BC069812.1 | - | 1 | 343 | 7262453 | ||
CB996360.1 | - | 2 | 549 | |||
M76673.1 | - | 1 | 343 |
Set Membership: |
G-protein coupled receptor |