Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145853.1 | NP_001139325.1 | 5 | 713 | ||
NM_006005.3 | NP_005996.2 | 5 | 717 | |||
XM_017008586.1 | XP_016864075.1 | 6 | 696 | |||
GenBank mRNA | AF084481.1 | - | 5 | 717 | ||
AK312897.1 | - | 5 | 646 | |||
BC030130.2 | - | 5 | 647 | 4127654 | ||
KU178395.1 | - | 4 | 547 | |||
Y18064.1 | - | 5 | 705 |