Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318051.1 | NP_001304980.1 | 4 | 470 | ||
NM_002495.3 | NP_002486.1 | 5 | 544 | |||
NR_134473.1 | 6 | 716 | ||||
NR_134474.1 | 6 | 633 | ||||
NR_134475.1 | 6 | 668 | ||||
GenBank mRNA | AF020351.1 | - | 5 | 522 | ||
AK307973.1 | - | 5 | 521 | |||
BC005270.1 | - | 5 | 538 | 3681312 | ||
BU145342.1 | - | 4 | 444 | |||
BU954271.1 | - | 5 | 536 | |||
CD692133.1 | - | 6 | 684 | |||
KU178226.1 | - | 5 | 514 | |||
KU178227.1 | - | 6 | 686 |