Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134473.1 | 3 | 317 | |||
NR_134474.1 | 3 | 234 | ||||
XM_017009491.1 | XP_016864980.1 | 3 | 340 | |||
GenBank mRNA | DA609203.1 | - | 3 | 234 | ||
KU178227.1 | - | 3 | 287 |