Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 20, 21 | 4788 | ||
NM_018136.4 | NP_060606.3 | 21, 22 | 9543 | |||
GenBank mRNA | AF509326.1 | - | 21, 22 | 9286 | ||
AK001379.1 | - | 4, 5 | 2677 | |||
AK001411.1 | - | 4, 5 | 1070 | |||
AK226178.1 | - | 20, 21 | 4720 | |||
AY367065.1 | - | 21, 22 | 9286 | |||
AY971955.1 | - | 7, 8 | 2041 | |||
AY971956.1 | - | 20, 21 | 4531 | |||
AY971957.1 | - | 7, 8 | 3244 | |||
BC034607.1 | - | 16, 17 | 2407 | |||
BC040439.1 | - | 4, 5 | 1105 | |||
BX648804.1 | - | 4, 5 | 4397 |