Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206846.1 | NP_001193775.1 | 23 | 5232 | ||
NM_018136.4 | NP_060606.3 | 24 | 9987 | |||
GenBank mRNA | AF509326.1 | - | 24 | 9730 | ||
AK001379.1 | - | 7 | 3121 | |||
AK001411.1 | - | 7 | 1514 | |||
AK226178.1 | - | 23 | 5164 | |||
AY367065.1 | - | 24 | 9730 | |||
AY971955.1 | - | 10 | 2485 | |||
AY971956.1 | - | 23 | 4975 | |||
AY971957.1 | - | 10 | 3688 | |||
BC034607.1 | - | 19 | 2851 | |||
BC040439.1 | - | 7 | 1549 | |||
BX648804.1 | - | 7 | 4841 |