Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000017.3 | NP_000008.1 | 10 | 1921 | ||
NM_001302554.1 | NP_001289483.1 | 10 | 1909 | |||
GenBank mRNA | M26393.1 | - | 10 | 1804 |