Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001790.2 | NP_001001790.1 | 2 | 686 | ||
NM_001134484.1 | NP_001127956.1 | 2 | 845 | |||
NM_001134485.1 | NP_001127957.1 | 2 | 788 | |||
GenBank mRNA | BC034247.1 | - | 2 | 618 | 3861303 |