Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033054.2 | NP_001028226.1 | 4 | 672 | ||
NM_001033055.2 | NP_001028227.1 | 4 | 681 | |||
NM_001285399.2 | NP_001272328.1 | 5 | 825 | |||
NM_001285400.2 | NP_001272329.1 | 5 | 795 | |||
NM_001285401.2 | NP_001272330.1 | 5 | 789 | |||
NM_001285402.1 | NP_001272331.1 | 5 | 852 | |||
NM_001285403.2 | NP_001272332.1 | 5 | 837 | |||
NM_014336.4 | NP_055151.3 | 5 | 861 | |||
GenBank mRNA | AB593042.1 | - | 5 | 758 | ||
AB593043.1 | - | 5 | 782 | |||
AB593044.1 | - | 5 | 796 | |||
AB593045.1 | - | 5 | 796 | |||
AB593046.1 | - | 5 | 798 | |||
AB593047.1 | - | 5 | 839 | |||
AB593048.1 | - | 5 | 852 | |||
AB593049.1 | - | 5 | 806 | |||
AB593050.1 | - | 5 | 782 | |||
AB593051.1 | - | 5 | 783 | |||
AB593052.1 | - | 5 | 782 | |||
AB593053.1 | - | 4 | 638 | |||
AB593054.1 | - | 4 | 581 | |||
AB593055.1 | - | 5 | 827 | |||
AF038437.1 | - | 5 | 780 | |||
AF148864.1 | - | 5 | 946 | |||
AF525970.1 | - | 4 | 672 | |||
AJ633677.1 | - | 4 | 634 | |||
AJ633678.1 | - | 4 | 625 | |||
AJ830742.1 | - | 5 | 749 | |||
AJ830743.1 | - | 5 | 743 | |||
AK023970.1 | - | 5 | 789 | |||
AK314558.1 | - | 5 | 782 | |||
BC012055.1 | - | 5 | 774 | 4634156 | ||
BM685943.1 | - | 1 | 59 | |||
BX537907.1 | - | 5 | 759 | |||
BX647279.1 | - | 5 | 827 |