Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000356.3 | NP_000347.2 | 20 | 3247 | ||
NM_001135243.1 | NP_001128715.1 | 21 | 3478 | |||
NM_001135244.1 | NP_001128716.1 | 20 | 3364 | |||
NM_001135245.1 | NP_001128717.1 | 20 | 3247 | |||
NM_001195141.1 | NP_001182070.1 | 20 | 3364 | |||
XM_005268502.3 | XP_005268559.1 | 22 | 3526 | |||
XM_005268503.3 | XP_005268560.1 | 22 | 3523 | |||
XM_005268504.3 | XP_005268561.1 | 22 | 3526 | |||
XM_005268505.3 | XP_005268562.1 | 21 | 3415 | |||
XM_005268506.3 | XP_005268563.1 | 21 | 3412 | |||
XM_005268507.3 | XP_005268564.1 | 21 | 3295 | |||
XM_011537678.2 | XP_011535980.1 | 21 | 3346 | |||
XM_017009792.1 | XP_016865281.1 | 21 | 3412 | |||
XM_017009793.1 | XP_016865282.1 | 20 | 3235 | |||
XM_017009794.1 | XP_016865283.1 | 19 | 3121 | |||
GenBank mRNA | AB209317.1 | - | 13 | 2308 | ||
AK096467.1 | - | 17 | 2983 | |||
AK299167.1 | - | 13 | 2321 | |||
AK303611.1 | - | 20 | 3289 | |||
AK307586.1 | - | 21 | 3403 | |||
AY460334.1 | - | 21 | 3370 | |||
BC008331.1 | - | 12 | 2044 | |||
BC016144.1 | - | 20 | 3284 | |||
BC027252.1 | - | 20 | 3095 | |||
U40847.1 | - | 20 | 3232 | |||
U76366.1 | - | 20 | 3214 |