Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008844.2 | NP_001008844.1 | 23 | 3867 | ||
NM_001319034.1 | NP_001305963.1 | 23 | 3867 | |||
NM_004415.3 | NP_004406.2 | 23 | 3867 | |||
GenBank mRNA | AB209992.1 | - | 23 | 3825 | ||
AK304182.1 | - | 23 | 3770 | |||
BC140802.1 | - | 23 | 3800 | 9021670 | ||
HM151899.1 | - | 23 | 3805 | |||
J05211.1 | - | 1 | 169 | |||
M77830.3 | - | 23 | 3805 |