Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000192.3 | NP_000183.2 | 6, 7 | 1337 | ||
NM_080717.2 | NP_542448.1 | 5, 6 | 1152 | |||
NM_181486.2 | NP_852259.1 | 6, 7 | 1244 | |||
XM_017019912.1 | XP_016875401.1 | 6, 7 | 3088 | |||
GenBank mRNA | AB051068.1 | - | 5, 6 | 1097 | ||
AF221714.1 | - | 6, 7 | 1335 | |||
AK290611.1 | - | 6, 7 | 841 | |||
AK300251.1 | - | 4, 5 | 638 | |||
BC027942.1 | - | 6, 7 | 1165 | 5204163 | ||
U80987.1 | - | 5, 6 | 670 | |||
U89353.1 | - | 5, 6 | 670 | |||
Y09445.1 | - | 6, 7 | 1326 |