Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLA2G6 SIRNA Assays ›
Gene Symbol:
PLA2G6
Gene Name:
phospholipase A2 group VI
Gene Aliases:
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Chromosome Location:
Chr.22: 38111495 - 38192099 on Build GRCh38
UniGene ID:
Hs.170479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s536326
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLA2G6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004426.1
NP_001004426.1
6
950
NM_001199562.1
NP_001186491.1
6
905
NM_003560.2
NP_003551.2
6
950
XM_005261764.2
XP_005261821.1
6
925
XM_005261765.1
XP_005261822.1
6
922
XM_005261766.1
XP_005261823.1
6
1017
XM_006724332.3
XP_006724395.1
6
926
XM_011530422.1
XP_011528724.1
6
801
XM_011530423.1
XP_011528725.1
7
1037
XM_011530424.1
XP_011528726.1
8
1161
XM_011530425.1
XP_011528727.1
6
861
XM_011530426.2
XP_011528728.1
6
925
XM_017028981.1
XP_016884470.1
6
1017
XM_017028982.1
XP_016884471.1
6
922
XM_017028983.1
XP_016884472.1
8
1161
XM_017028984.1
XP_016884473.1
7
1037
XM_017028985.1
XP_016884474.1
5
693
XM_017028986.1
XP_016884475.1
6
925
XM_017028987.1
XP_016884476.1
6
925
XM_017028988.1
XP_016884477.1
6
925
GenBank mRNA
AF064594.1
-
6
950
AK001290.1
-
239
AK096521.1
-
5
483
AK290825.1
-
6
907
AK291212.1
-
6
936
AK300459.1
-
6
801
AK308308.1
-
5
756
AL080187.1
-
6
950
BC036742.1
-
6
944
5166749
BC051904.1
-
6
886
5175768
CR456543.1
-
6
878
Back To Top
More Information
Panther Classification:
Molecular Function -
phospholipase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
chemotaxis
positive regulation of cytosolic calcium ion concentration
memory
urinary bladder smooth muscle contraction
lipid catabolic process
antibacterial humoral response
cardiolipin biosynthetic process
response to endoplasmic reticulum stress
positive regulation of insulin secretion involved in cellular response to glucose stimulus
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of vasodilation
positive regulation of exocytosis
negative regulation of synaptic transmission, glutamatergic
maternal process involved in female pregnancy
positive regulation of protein kinase C signaling
positive regulation of release of cytochrome c from mitochondria
positive regulation of arachidonic acid secretion
regulation of store-operated calcium channel activity
positive regulation of ceramide biosynthetic process
phospholipase A2 activity
calmodulin binding
hydrolase activity
protein kinase binding
ATP-dependent protein binding
calcium-independent phospholipase A2 activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline