Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012267.2 | NP_001012267.1 | 7 | 1204 | ||
NM_001286969.1 | NP_001273898.1 | 6 | 1022 | |||
NM_001286971.1 | NP_001273900.1 | 3 | 361 | |||
GenBank mRNA | AK091247.1 | - | 3 | 362 | ||
AK092603.1 | - | 7 | 1019 | |||
AK307262.1 | - | 3 | 244 | |||
AK307435.1 | - | 6 | 542 | |||
AL833701.1 | - | 2 | 879 | |||
BC071726.1 | - | 7 | 730 | 5554572 | ||
BX537851.1 | - | 6 | 551 |