Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 10 | 962 | ||
NM_007297.3 | NP_009228.2 | 9 | 870 | |||
NM_007298.3 | NP_009229.2 | 9 | 749 | |||
NM_007299.3 | NP_009230.2 | 10 | 924 | |||
NM_007300.3 | NP_009231.2 | 10 | 962 | |||
NR_027676.1 | 10 | 866 | ||||
GenBank mRNA | AF005068.1 | - | 10 | 831 | ||
AK307553.1 | - | 9 | 765 | |||
AK308084.1 | - | 10 | 891 | |||
AK316200.1 | - | 9 | 821 | |||
AY354539.2 | - | 9 | 794 | |||
AY751490.1 | - | 9 | 730 | |||
BC030969.1 | - | 10 | 849 | |||
BC046142.1 | - | 4 | 355 | |||
BC062429.1 | - | 2 | 84 | |||
BC072418.1 | - | 10 | 924 | |||
BC085615.1 | - | 10 | 829 | |||
BC106745.1 | - | 10 | 779 | |||
BC106746.1 | - | 2 | 90 | |||
BC114511.1 | - | 11 | 895 | |||
BC115037.1 | - | 10 | 867 | 7961446 | ||
JN686490.1 | - | 9 | 730 | |||
U14680.1 | - | 10 | 849 | |||
U64805.1 | - | 9 | 730 |