Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 8 | 1001 | ||
XM_005264637.3 | XP_005264694.1 | 7 | 809 | |||
GenBank mRNA | AK057222.1 | - | 8 | 997 | ||
AK289953.1 | - | 8 | 984 | |||
AL833892.1 | - | 7 | 735 | |||
BC009488.2 | - | 7 | 724 |