Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145038.4 | NP_659475.2 | 8, 9 | 1114 | ||
XM_005264637.3 | XP_005264694.1 | 7, 8 | 922 | |||
GenBank mRNA | AK057222.1 | - | 8, 9 | 1110 | ||
AK289953.1 | - | 8, 9 | 1097 | |||
AL833892.1 | - | 7, 8 | 848 | |||
BC009488.2 | - | 7, 8 | 837 |