Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRP5 SIRNA Assays ›
Gene Symbol:
LRP5
Gene Name:
LDL receptor related protein 5
Gene Aliases:
BMND1, EVR1, EVR4, HBM, LR3, LRP-5, LRP7, OPPG, OPS, OPTA1, VBCH2
Chromosome Location:
Chr.11: 68298866 - 68449275 on Build GRCh38
UniGene ID:
Hs.6347
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s536382
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRP5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017017736.1
XP_016873225.1
1
89
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
positive regulation of mesenchymal cell proliferation
osteoblast development
glucose catabolic process
endocytosis
cholesterol metabolic process
regulation of blood pressure
positive regulation of cell proliferation
anterior/posterior pattern specification
Wnt signaling pathway
positive regulation of osteoblast proliferation
somatic stem cell population maintenance
extracellular matrix-cell signaling
cell migration involved in gastrulation
cholesterol homeostasis
embryonic digit morphogenesis
regulation of apoptotic process
Wnt signaling pathway involved in dorsal/ventral axis specification
positive regulation of fat cell differentiation
negative regulation of osteoblast differentiation
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
bone remodeling
regulation of bone remodeling
bone marrow development
positive regulation of sequence-specific DNA binding transcription factor activity
anatomical structure regression
retina morphogenesis in camera-type eye
canonical Wnt signaling pathway
bone morphogenesis
branching involved in mammary gland duct morphogenesis
adipose tissue development
cell-cell signaling involved in mammary gland development
regulation of canonical Wnt signaling pathway
regulation of insulin secretion involved in cellular response to glucose stimulus
retinal blood vessel morphogenesis
negative regulation of protein serine/threonine kinase activity
apoptotic process involved in patterning of blood vessels
beta-catenin destruction complex disassembly
protein binding
Wnt-protein binding
toxin transporter activity
Wnt-activated receptor activity
coreceptor activity involved in Wnt signaling pathway
coreceptor activity involved in canonical Wnt signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline