Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 15 | 2254 | ||
NM_001320665.1 | NP_001307594.1 | 15 | 2232 | |||
NM_033028.4 | NP_149017.2 | 16 | 2301 | |||
NR_045565.1 | 17 | 2359 | ||||
NR_045566.1 | 16 | 2614 | ||||
XM_011521848.1 | XP_011520150.1 | 16 | 2276 | |||
XM_011521849.1 | XP_011520151.1 | 15 | 2164 | |||
XM_011521851.1 | XP_011520153.1 | 14 | 2152 | |||
XM_017022450.1 | XP_016877939.1 | 15 | 2513 | |||
XM_017022451.1 | XP_016877940.1 | 15 | 2242 | |||
XM_017022452.1 | XP_016877941.1 | 15 | 2472 | |||
XM_017022453.1 | XP_016877942.1 | 15 | 2159 | |||
XM_017022454.1 | XP_016877943.1 | 14 | 2109 | |||
GenBank mRNA | AF359281.1 | - | 16 | 2273 | ||
AY457143.1 | - | 16 | 2373 | |||
BC027624.2 | - | 16 | 2271 | 4824276 |