Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252678.1 | NP_001239607.1 | 15 | 1849 | ||
NM_001320665.1 | NP_001307594.1 | 15 | 1827 | |||
NM_033028.4 | NP_149017.2 | 16 | 1896 | |||
NR_045565.1 | 17 | 1954 | ||||
NR_045566.1 | 16 | 2209 | ||||
XM_011521848.1 | XP_011520150.1 | 16 | 1871 | |||
XM_011521849.1 | XP_011520151.1 | 15 | 1759 | |||
XM_011521851.1 | XP_011520153.1 | 14 | 1747 | |||
XM_017022450.1 | XP_016877939.1 | 15 | 2108 | |||
XM_017022451.1 | XP_016877940.1 | 15 | 1837 | |||
XM_017022452.1 | XP_016877941.1 | 15 | 2067 | |||
XM_017022453.1 | XP_016877942.1 | 15 | 1754 | |||
XM_017022454.1 | XP_016877943.1 | 14 | 1704 | |||
GenBank mRNA | AF359281.1 | - | 16 | 1868 | ||
AK075321.1 | - | 16 | 1860 | |||
AK303706.1 | - | 15 | 1844 | |||
AY457143.1 | - | 16 | 1968 | |||
BC008923.2 | - | 13 | 1673 | |||
BC027624.2 | - | 16 | 1866 | 4824276 | ||
BX647855.1 | - | 15 | 1820 |