Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126131.1 | NP_001119603.1 | 4 | 1199 | ||
NM_002693.2 | NP_002684.1 | 4 | 1211 | |||
GenBank mRNA | AK291281.1 | - | 4 | 1230 | ||
BC042571.1 | - | 4 | 1196 | 4384475 | ||
BC050559.1 | - | 4 | 1180 | 6042595 | ||
D84103.1 | - | 3 | 1035 | |||
U60325.1 | - | 3 | 1088 | |||
X98093.1 | - | 4 | 1211 |