Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000547.5 | NP_000538.3 | 12 | 2178 | ||
NM_001206744.1 | NP_001193673.1 | 12 | 2173 | |||
NM_001206745.1 | NP_001193674.1 | 11 | 2002 | |||
NM_175719.3 | NP_783650.1 | 11 | 2007 | |||
NM_175721.3 | NP_783652.1 | 11 | 2088 | |||
NM_175722.3 | NP_783653.1 | 10 | 1569 | |||
XM_011510379.2 | XP_011508681.1 | 11 | 2088 | |||
XM_011510380.2 | XP_011508682.1 | 11 | 2088 | |||
XM_011510381.2 | XP_011508683.1 | 10 | 1917 | |||
GenBank mRNA | AB208960.1 | - | 9 | 1874 | ||
AF439430.1 | - | 10 | 1568 | |||
AF533530.1 | - | 3 | 511 | |||
AF533531.1 | - | 3 | 509 | |||
AY136822.1 | - | 1 | 25 | |||
BC095448.1 | - | 12 | 2171 | 30329791 | ||
J02969.1 | - | 12 | 2159 | |||
J02970.1 | - | 11 | 1988 | |||
M17755.2 | - | 12 | 2171 | |||
M55702.1 | - | 1 | 80 | |||
X17358.1 | - | 11 | 1981 | |||
Y00406.1 | - | 12 | 2127 |