Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042355.1 | NP_001035814.1 | 18 | 5877 | ||
NM_001042412.1 | NP_001035877.1 | 18 | 5901 | |||
NM_015247.2 | NP_056062.1 | 20 | 6023 | |||
XM_005255812.2 | XP_005255869.1 | 19 | 5974 | |||
XM_006721149.1 | XP_006721212.1 | 19 | 6038 | |||
XM_011522907.2 | XP_011521209.1 | 18 | 5800 | |||
XM_011522908.1 | XP_011521210.1 | 18 | 5862 | |||
XM_017022977.1 | XP_016878466.1 | 18 | 6172 | |||
XM_017022978.1 | XP_016878467.1 | 18 | 5968 | |||
XM_017022979.1 | XP_016878468.1 | 18 | 5928 | |||
XM_017022980.1 | XP_016878469.1 | 18 | 5961 | |||
XM_017022981.1 | XP_016878470.1 | 18 | 5829 | |||
GenBank mRNA | DA940312.1 | - | 1 | 245 |