Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014080.4 | NP_054799.4 | 21, 22 | 3057 | ||
XM_005254421.2 | XP_005254478.1 | 21, 22 | 3057 | |||
GenBank mRNA | AF181972.1 | - | 13, 14 | 1840 | ||
AF230496.1 | - | 21, 22 | 3039 | |||
AF267981.1 | - | 21, 22 | 3057 |