Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006915.2 | NP_008846.2 | 2 | 449 | ||
GenBank mRNA | AJ007590.1 | - | 2 | 432 | ||
AK312555.1 | - | 2 | 283 | |||
BC043348.2 | - | 2 | 371 | 5520795 | ||
BC053530.1 | - | 2 | 369 | 3871941 | ||
DA211161.1 | - | 2 | 450 |