Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000350.2 | NP_000341.2 | 4, 5 | 551 | ||
GenBank mRNA | AB210040.1 | - | 4, 5 | 537 | ||
AF000148.1 | - | 4, 5 | 534 | |||
AF001945.1 | - | 4, 5 | 528 | |||
AK295818.1 | - | 4, 5 | 527 | |||
U88667.1 | - | 4, 5 | 528 |