Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 15 | 1988 | ||
XM_005274604.2 | XP_005274661.1 | 14 | 1868 | |||
XM_005274606.3 | XP_005274663.1 | 16 | 2130 | |||
XM_011545592.2 | XP_011543894.1 | 16 | 1959 | |||
XM_011545594.2 | XP_011543896.1 | 14 | 1409 | |||
XM_011545595.1 | XP_011543897.1 | 14 | 1419 | |||
XM_011545597.1 | XP_011543899.1 | 16 | 2044 | |||
XM_011545598.1 | XP_011543900.1 | 4 | 487 | |||
XM_017029909.1 | XP_016885398.1 | 15 | 1564 | |||
XM_017029910.1 | XP_016885399.1 | 16 | 1772 | |||
XM_017029911.1 | XP_016885400.1 | 7 | 809 | |||
GenBank mRNA | AK225847.1 | - | 14 | 1798 | ||
AK290354.1 | - | 15 | 1973 | |||
AK297104.1 | - | 7 | 768 | |||
BC096344.3 | - | 14 | 1683 | 40008704 | ||
GU727634.1 | - | 14 | 1629 | |||
Y15164.1 | - | 15 | 1940 | |||
Y16355.1 | - | 15 | 2604 |