Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003611.2 | NP_003602.1 | 16 | 2065 | ||
XM_005274604.2 | XP_005274661.1 | 15 | 1945 | |||
XM_005274606.3 | XP_005274663.1 | 17 | 2207 | |||
XM_011545592.2 | XP_011543894.1 | 17 | 2036 | |||
XM_011545594.2 | XP_011543896.1 | 15 | 1486 | |||
XM_011545595.1 | XP_011543897.1 | 15 | 1496 | |||
XM_011545597.1 | XP_011543899.1 | 17 | 2121 | |||
XM_011545598.1 | XP_011543900.1 | 5 | 564 | |||
XM_017029909.1 | XP_016885398.1 | 16 | 1641 | |||
XM_017029910.1 | XP_016885399.1 | 17 | 1849 | |||
XM_017029911.1 | XP_016885400.1 | 8 | 886 | |||
GenBank mRNA | AK225847.1 | - | 15 | 1875 | ||
AK290354.1 | - | 16 | 2050 | |||
AK297104.1 | - | 8 | 845 | |||
BC096344.3 | - | 15 | 1760 | 40008704 | ||
BC096345.1 | - | 15 | 1774 | |||
GU727634.1 | - | 15 | 1706 | |||
Y15164.1 | - | 16 | 2017 | |||
Y16355.1 | - | 16 | 2681 |