Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s536528
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
4
517
NM_001122764.1
NP_001116236.1
4
578
XM_005245291.3
XP_005245348.2
4
556
XM_006711402.2
XP_006711465.2
4
523
XM_006711403.2
XP_006711466.2
4
512
XM_006711404.3
XP_006711467.2
4
460
XM_006711406.2
XP_006711469.2
4
414
XM_011509663.2
XP_011507965.1
4
643
XM_011509664.1
XP_011507966.1
4
414
XM_011509665.2
XP_011507967.1
4
641
XM_011509666.2
XP_011507968.1
4
648
XM_011509667.2
XP_011507969.1
4
584
XM_011509668.2
XP_011507970.1
4
384
XM_011509669.1
XP_011507971.1
4
397
XM_011509670.2
XP_011507972.1
4
646
XM_011509671.1
XP_011507973.1
4
414
XM_011509672.2
XP_011507974.1
4
637
XM_011509674.2
XP_011507976.1
4
649
XM_011509675.2
XP_011507977.1
4
579
XM_011509676.1
XP_011507978.1
3
709
XM_011509677.1
XP_011507979.1
3
642
XM_011509678.1
XP_011507980.1
4
602
XM_011509679.1
XP_011507981.1
4
410
XM_011509681.1
XP_011507983.1
4
460
XM_011509682.1
XP_011507984.1
4
501
XM_017001559.1
XP_016857048.1
4
640
XM_017001560.1
XP_016857049.1
4
556
XM_017001561.1
XP_016857050.1
4
541
XM_017001562.1
XP_016857051.1
4
483
XM_017001563.1
XP_016857052.1
4
600
XM_017001565.1
XP_016857054.1
4
483
XM_017001566.1
XP_016857055.1
4
592
XM_017001568.1
XP_016857057.1
4
592
XM_017001569.1
XP_016857058.1
4
460
GenBank mRNA
AK094855.1
-
4
464
AK302298.1
-
4
502
BC002357.2
-
4
498
2821983
BX384814.2
-
4
592
D38537.1
-
4
565
U26446.1
-
4
479
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9xzrh:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline