Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039550.1 | NP_001034639.1 | 7 | 738 | ||
NM_006736.5 | NP_006727.2 | 7 | 738 | |||
GenBank mRNA | AK289626.1 | - | 7 | 561 | ||
AK292761.1 | - | 7 | 566 | |||
AK296297.1 | - | 2 | 288 | |||
AK312723.1 | - | 7 | 745 | |||
AL547521.3 | - | 7 | 784 | |||
BC011609.2 | - | 7 | 680 | 3619734 | ||
BC047056.1 | - | 6 | 541 | 5185009 | ||
BT007088.1 | - | 6 | 501 | |||
S37374.1 | - | 6 | 526 | |||
S37375.1 | - | 6 | 526 |