Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039550.1 | NP_001034639.1 | 10 | 1095 | ||
NM_006736.5 | NP_006727.2 | 9 | 2246 | |||
GenBank mRNA | BC011609.2 | - | 9 | 2188 | 3619734 | |
BC047056.1 | - | 9 | 898 | 5185009 | ||
BF795662.1 | - | 1 | 397 | |||
S37374.1 | - | 9 | 2002 | |||
S37375.1 | - | 9 | 883 |