Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002292.3 | NP_002283.3 | 24 | 3950 | ||
XM_005265127.3 | XP_005265184.1 | 25 | 3749 | |||
GenBank mRNA | AK094050.1 | - | 5 | 569 | ||
CV814167.1 | - | 4 | 689 | |||
S77512.1 | - | 24 | 3803 | |||
X72760.1 | - | 1 | 105 | |||
X79683.1 | - | 25 | 3816 |