Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033380.1 | 6 | 541 | |||
NR_033381.1 | 6 | 541 | ||||
GenBank mRNA | AK023533.1 | - | 2 | 2179 | ||
AK125149.1 | - | 6 | 414 | |||
BC019893.1 | - | 6 | 412 |