Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000432.3 | NP_000423.2 | 4 | 301 | ||
GenBank mRNA | AF020768.1 | - | 3 | 235 | ||
AK312013.1 | - | 4 | 278 | |||
BC015821.1 | - | 4 | 283 | 4245364 | ||
BC031006.1 | - | 4 | 278 | 4292208 | ||
BC031008.1 | - | 4 | 280 | 4297454 | ||
CR456962.1 | - | 3 | 230 | |||
CR541957.1 | - | 3 | 230 | |||
S69022.1 | - | 4 | 307 | |||
X14332.1 | - | 4 | 227 | |||
X66141.1 | - | 4 | 260 |