Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 10 | 2378 | ||
NM_001306179.1 | NP_001293108.1 | 10 | 2399 | |||
XM_005253931.3 | XP_005253988.1 | 9 | 2524 | |||
GenBank mRNA | HM116542.1 | - | 10 | 1994 | ||
HM116543.1 | - | 8 | 1783 | |||
HM116544.1 | - | 7 | 1437 | |||
HM116545.1 | - | 7 | 1307 | |||
HM116546.1 | - | 5 | 937 | |||
HM116549.1 | - | 4 | 637 | |||
HM116551.1 | - | 2 | 493 | |||
HM449088.1 | - | 9 | 2293 | |||
HM449089.1 | - | 9 | 2000 | |||
M57732.1 | - | 10 | 2200 |