Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000545.6 | NP_000536.5 | 10 | 2723 | ||
NM_001306179.1 | NP_001293108.1 | 10 | 2744 | |||
XM_005253931.3 | XP_005253988.1 | 9 | 2869 | |||
GenBank mRNA | BM854591.1 | - | 1 | 79 | ||
HM116541.1 | - | 11 | 1980 | |||
HM116542.1 | - | 10 | 2339 | |||
HM116543.1 | - | 8 | 2128 | |||
HM116544.1 | - | 7 | 1782 | |||
HM116545.1 | - | 7 | 1652 | |||
HM116546.1 | - | 5 | 1282 | |||
HM116549.1 | - | 4 | 982 | |||
HM116550.1 | - | 2 | 400 | |||
HM116551.1 | - | 2 | 838 | |||
HM449088.1 | - | 9 | 2638 | |||
HM449089.1 | - | 9 | 2345 | |||
M57732.1 | - | 10 | 2546 |