Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000243.2 | NP_000234.1 | 10 | 1947 | ||
NM_001198536.1 | NP_001185465.1 | 9 | 1489 | |||
XM_017023236.1 | XP_016878725.1 | 10 | 2085 | |||
GenBank mRNA | AF018080.1 | - | 10 | 1948 | ||
AK298660.1 | - | 10 | 1941 | |||
BC101511.1 | - | 10 | 1932 | 8069017 | ||
BC101537.1 | - | 10 | 1932 | 8069043 | ||
FJ785717.1 | - | 7 | 828 | |||
FJ785718.1 | - | 6 | 712 | |||
FJ785719.1 | - | 5 | 679 | |||
FJ785720.1 | - | 6 | 597 | |||
FJ785721.1 | - | 6 | 1312 | |||
FJ785722.1 | - | 9 | 1451 | |||
Y14441.1 | - | 10 | 1936 |