Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000243.2 | NP_000234.1 | 4, 5 | 1389 | ||
NM_001198536.1 | NP_001185465.1 | 3, 4 | 756 | |||
XM_017023236.1 | XP_016878725.1 | 4, 5 | 1527 | |||
GenBank mRNA | AF018080.1 | - | 4, 5 | 1390 | ||
AK298660.1 | - | 4, 5 | 1383 | |||
BC101511.1 | - | 4, 5 | 1374 | 8069017 | ||
BC101537.1 | - | 4, 5 | 1374 | 8069043 | ||
FJ785722.1 | - | 3, 4 | 716 | |||
Y14441.1 | - | 4, 5 | 1378 |